Learn More
Santa Cruz Biotechnology Anti-SREBP-1 Antibody (2A4), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC13551
Encompass
Anti-SREBP-1 Antibody (2A4) is recommended for detection of SREBP-1 of mouse, rat, human and hamster origin by WB, IP, IF and IHC(P).
Catalog No. NC9270469