Learn More
Cayman Chemical SREBP2 Transcription Factor A
SDP
Catalog No. 502886782
Encompass_Preferred
SREBP-2 is a transcription factor that performs a critical role in the transcriptional regulation of genes involved in cholesterol synthesis and uptake including HMG-CoA synthase, HMG-CoA reductase, and the LDL receptor.
Catalog No. 50-288-6782 Supplier Cayman Chemical Supplier No. 1000781996 WELL
SREBP-2 is a transcription factor that performs a critical role in the transcriptional regulation of genes involved in cholesterol synthesis and uptake including HMG-CoA synthase, HMG-CoA reductase, and the LDL receptor.