Learn More
ABclonal Technology SREK1 Rabbit pAb
SDP
Catalog No. 5000097910
Encompass_Preferred
This gene encodes a member of a family of serine/arginine-rich (SR) splicing proteins containing RNA recognition motif (RRM) domains. The encoded protein interacts with other SR proteins to modulate splice site selection. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Catalog No. 50-000-97910 Supplier ABclonal Technology Supplier No. A132351000UL
This gene encodes a member of a family of serine/arginine-rich (SR) splicing proteins containing RNA recognition motif (RRM) domains. The encoded protein interacts with other SR proteins to modulate splice site selection. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.