Learn More
Abeomics Srf-Re Reporter 293 Cell Line
Catalog No. NC1745350
Encompass_Preferred
The SRF-RE Leeporter Luciferase Reporter cell line is a stably transfected HEK 293 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the serum response factor-response element (SRF-RE). The SRF-RE reporter cell line can be used for studying GPCR-linked RhoA signaling pathways as well as screening of agonists, antagonists or signaling inhibitors related with the RhoA signaling pathways. Functional activity of the cell line has been validated by serum stimulation assay. Cell line application: (1) Monitor the GPCR-linked RhoA signaling pathway, and (2) Screen for activators or inhibitors of the GPCR-linked RhoA signaling pathway. .
Catalog No. NC1745350 Supplier Abeomics Supplier No. 14133ACL
The SRF-RE Leeporter Luciferase Reporter cell line is a stably transfected HEK 293 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the serum response factor-response element (SRF-RE). The SRF-RE reporter cell line can be used for studying GPCR-linked RhoA signaling pathways as well as screening of agonists, antagonists or signaling inhibitors related with the RhoA signaling pathways. Functional activity of the cell line has been validated by serum stimulation assay. Cell line application: (1) Monitor the GPCR-linked RhoA signaling pathway, and (2) Screen for activators or inhibitors of the GPCR-linked RhoA signaling pathway. .