Learn More
Aviva Systems Biology Corp Srgap1 Antibody - middle region (ARP57446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Srgap1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Arhgap13, 4930572H05Rik, Srgap1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-312