Learn More
Aviva Systems Biology Corp SRGN Antibody - middle region (ARP40416_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40416_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRGN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SRGN. Alternate names: FLJ12930, MGC9289, PPG, PRG, PRG1ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57437