Learn More
Aviva Systems Biology Corp SRGN Antibody - N-terminal region (ARP40415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRGN. It was validated on Western Blot. . Alternate names: FLJ12930, MGC9289, PPG, PRG, PRG1ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-284-870