Learn More
Medchemexpress LLC SRI-011381 | 1629138-41-5 | 99.6% | 329.48 | C20H31N3O | 10 MG
SDP
Catalog No. 5000214113
Encompass_Preferred
SRI-011381 is a small-molecule agonist of the TGF-β/Smad signaling pathway, reported to be orally active and to exhibit neuroprotective effects in preclinical studies. It is supplied as a solid research reagent with high reported purity and supporting documentation.
  • Orally active TGF-β/Smad signaling agonist.
  • Reported neuroprotective effects in preclinical models.
  • High purity suitable for research applications.
  • Supplied as a solid; hydrochloride salt is also reported as an alternate form.
  • Documentation available: datasheet, COA, and SDS.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-14113 Supplier Medchemexpress LLC Supplier No. HY10034710MG
SRI-011381 is a small-molecule agonist of the TGF-β/Smad signaling pathway, reported to be orally active and to exhibit neuroprotective effects in preclinical studies. It is supplied as a solid research reagent with high reported purity and supporting documentation.
  • Orally active TGF-β/Smad signaling agonist.
  • Reported neuroprotective effects in preclinical models.
  • High purity suitable for research applications.
  • Supplied as a solid; hydrochloride salt is also reported as an alternate form.
  • Documentation available: datasheet, COA, and SDS.
  • Intended for research use only; not for human consumption.