Learn More
Medchemexpress LLC SRI-011381 | 1629138-41-5 | MFCD01475985 | 99.6% | 329.48 g/mol | C20H31N3O | 25 MG
SDP
Catalog No. 5000214357
Encompass_Preferred
SRI-011381 is a small-molecule research compound that acts as an orally active agonist of the TGF-β/Smad signaling pathway and has reported neuroprotective effects. It is supplied for research use only and is characterized by defined analytical identifiers and purity.
  • Orally active TGF-β/Smad pathway agonist with reported neuroprotective effects.
  • High purity, typically around 99.6%.
  • Available in multiple solid pack sizes and as a DMSO solution for assay use.
  • Characterized by CAS 1629138-41-5 and molecular formula C20H31N3O.
  • Intended for research use only; not for human use.
Catalog No. 50-002-14357 Supplier Medchemexpress LLC Supplier No. HY10034725MG
SRI-011381 is a small-molecule research compound that acts as an orally active agonist of the TGF-β/Smad signaling pathway and has reported neuroprotective effects. It is supplied for research use only and is characterized by defined analytical identifiers and purity.
  • Orally active TGF-β/Smad pathway agonist with reported neuroprotective effects.
  • High purity, typically around 99.6%.
  • Available in multiple solid pack sizes and as a DMSO solution for assay use.
  • Characterized by CAS 1629138-41-5 and molecular formula C20H31N3O.
  • Intended for research use only; not for human use.