Learn More
Medchemexpress LLC SRI-011381 hydrochloride | 2070014-88-7 | 99.6% | 365.94 | C20H32ClN3O | 200 MG
SDP
Catalog No. 5000214372
Encompass_Preferred
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist reported to exhibit neuroprotective effects in vitro and in vivo. It is supplied as a solid or as a DMSO solution for research use and is provided at high chemical purity.
  • Orally active TGF-β signaling agonist.
  • Promotes proliferation of mouse lung fibroblasts in vitro.
  • Increases TGF-β1, NALP3, collagen-1, and α-SMA expression.
  • Demonstrates neuroprotective effects in animal models, reducing inflammation and neuron loss.
  • Available as a solid and as 10 mM solution in DMSO in multiple package sizes.
  • High purity suitable for research applications.
Catalog No. 50-002-14372 Supplier Medchemexpress LLC Supplier No. HY100347A200MG
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist reported to exhibit neuroprotective effects in vitro and in vivo. It is supplied as a solid or as a DMSO solution for research use and is provided at high chemical purity.
  • Orally active TGF-β signaling agonist.
  • Promotes proliferation of mouse lung fibroblasts in vitro.
  • Increases TGF-β1, NALP3, collagen-1, and α-SMA expression.
  • Demonstrates neuroprotective effects in animal models, reducing inflammation and neuron loss.
  • Available as a solid and as 10 mM solution in DMSO in multiple package sizes.
  • High purity suitable for research applications.