Learn More
Medchemexpress LLC SRI-011381 hydrochloride | 2070014-88-7 | 99.6% | 365.94 g·mol⁻¹ | C20H32ClN3O | 1 ML
SDP
Catalog No. 5000214008
Encompass_Preferred
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist reported to exhibit neuroprotective effects. It is supplied for research use as a high-purity solid and as a 10 mM solution in DMSO suitable for in vitro and preclinical studies.
  • Orally active TGF-β signaling agonist.
  • Reported to exhibit neuroprotective effects.
  • Supplied as solid and as a 10 mM solution in DMSO.
  • High purity (99.55%).
  • Available in multiple small and bulk quantities.
  • For research use only; not for human consumption.
Catalog No. 50-002-14008 Supplier Medchemexpress LLC Supplier No. HY100347A10MM 1ML
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist reported to exhibit neuroprotective effects. It is supplied for research use as a high-purity solid and as a 10 mM solution in DMSO suitable for in vitro and preclinical studies.
  • Orally active TGF-β signaling agonist.
  • Reported to exhibit neuroprotective effects.
  • Supplied as solid and as a 10 mM solution in DMSO.
  • High purity (99.55%).
  • Available in multiple small and bulk quantities.
  • For research use only; not for human consumption.