Learn More
Medchemexpress LLC SRI-011381 hydrochloride | 2070014-88-7 | 99.6% | 365.94 g/mol | C20H32ClN3O | 25 MG
SDP
Catalog No. 5000214136
Encompass_Preferred
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist used as a research reagent to modulate TGF-β/Smad pathways and to investigate neuroprotective effects. It is supplied as a white to off-white solid and is intended for laboratory research applications.
  • Orally active TGF-β signaling agonist with reported neuroprotective effects.
  • High purity (99.6%).
  • Molecular weight 365.94 g/mol; formula C20H32ClN3O.
  • White to off-white solid physical form suitable for handling in lab settings.
  • Available in milligram to gram quantities for in vitro and preclinical research.
Catalog No. 50-002-14136 Supplier Medchemexpress LLC Supplier No. HY100347A25MG
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist used as a research reagent to modulate TGF-β/Smad pathways and to investigate neuroprotective effects. It is supplied as a white to off-white solid and is intended for laboratory research applications.
  • Orally active TGF-β signaling agonist with reported neuroprotective effects.
  • High purity (99.6%).
  • Molecular weight 365.94 g/mol; formula C20H32ClN3O.
  • White to off-white solid physical form suitable for handling in lab settings.
  • Available in milligram to gram quantities for in vitro and preclinical research.