Learn More
Medchemexpress LLC SRI-011381 hydrochloride | 2070014-88-7 | 99.6% | 365.94 g/mol | C20H32ClN3O | 50 MG
SDP
Catalog No. 5000214429
Encompass_Preferred
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist used in research to modulate TGF-β/Smad pathways and investigate neuroprotective mechanisms. It shows activity in cell-based assays and animal studies, including promotion of fibroblast proliferation and mitigation of inflammation-related neuronal loss.
  • Orally active TGF-β signaling agonist.
  • Demonstrates neuroprotective effects in vitro and in vivo.
  • Promotes fibroblast proliferation and increases TGF-β1, NALP3, collagen-1, and α-SMA expression.
  • Shown to reduce inflammatory infiltration and alleviate neuronal loss in animal models.
  • Supplied as a solid with high purity and defined storage recommendations for research use.
Catalog No. 50-002-14429 Supplier Medchemexpress LLC Supplier No. HY100347A50MG
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist used in research to modulate TGF-β/Smad pathways and investigate neuroprotective mechanisms. It shows activity in cell-based assays and animal studies, including promotion of fibroblast proliferation and mitigation of inflammation-related neuronal loss.
  • Orally active TGF-β signaling agonist.
  • Demonstrates neuroprotective effects in vitro and in vivo.
  • Promotes fibroblast proliferation and increases TGF-β1, NALP3, collagen-1, and α-SMA expression.
  • Shown to reduce inflammatory infiltration and alleviate neuronal loss in animal models.
  • Supplied as a solid with high purity and defined storage recommendations for research use.