Learn More
Medchemexpress LLC HY-123600 5mg Medchemexpress, SRI-29329 CAS:2086809-58-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1236005MG
Encompass_Preferred
Medchemexpress, HY-123600 5mg SRI-29329 CAS:2086809-58-5 SRI-29329 is a specific CLK inhibitor, with IC50 values of 78 nM, 16 nM and 86 nM for CLK1, CLK2 and CLK4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1020