Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SRI 31215 TFA 10mM 1mL | 1832686-44-8 | 1 ML
SDP

Catalog No. 5000229743
Encompass_Preferred

SRI 31215 (trifluoroacetate) is the TFA salt of a small-molecule inhibitor targeting matriptase, hepsin, and hepatocyte growth factor activator (HGFA). Supplied as a 10 mM solution in DMSO (1 mL), it is intended for in vitro research applications.

  • Trifluoroacetate salt.
  • Matriptase, hepsin, and HGFA inhibitor.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High reported purity of 98.5%.
  • Molecular formula C27H34F3N5O3; molecular weight 533.59 g/mol.

Catalog No. 50-002-29743 Supplier Medchemexpress LLC Supplier No. HY114363A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SRI 31215 (trifluoroacetate) is the TFA salt of a small-molecule inhibitor targeting matriptase, hepsin, and hepatocyte growth factor activator (HGFA). Supplied as a 10 mM solution in DMSO (1 mL), it is intended for in vitro research applications.

  • Trifluoroacetate salt.
  • Matriptase, hepsin, and HGFA inhibitor.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High reported purity of 98.5%.
  • Molecular formula C27H34F3N5O3; molecular weight 533.59 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.