Learn More
Medchemexpress LLC HY-114363A 5mg Medchemexpress, SRI 31215 (TFA) CAS:1832686-44-8 Purity:>98%
SDP
Catalog No. 501932482
Encompass_Preferred
Medchemexpress, HY-114363A 5mg SRI 31215 (TFA) CAS:1832686-44-8 SRI 31215 (TFA), a triplex inhibitor of matriptase , hepsin and hepatocyte growth factor activator (HGFA) with IC 50 s of 0.69 μM, 0.65 μM, 0.3 μM, blocks pro-HGF activation and thus mimics the activity of HAI-1/2 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2482 Supplier Medchemexpress LLC Supplier No. HY114363A5MG
Medchemexpress, HY-114363A 5mg SRI 31215 (TFA) CAS:1832686-44-8 SRI 31215 (TFA), a triplex inhibitor of matriptase , hepsin and hepatocyte growth factor activator (HGFA) with IC 50 s of 0.69 μM, 0.65 μM, 0.3 μM, blocks pro-HGF activation and thus mimics the activity of HAI-1/2 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.