Learn More
Medchemexpress LLC SRI-37330 | 2322245-42-9 | 99.2% | 388.41 | C16H19F3N4O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1421145MG
Encompass_Preferred
SRI-37330 is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor used in preclinical research. It has been shown to inhibit glucagon secretion and function, reduce hepatic glucose production, and reverse hepatic steatosis, making it useful for studies of type 2 diabetes and metabolic disease. The compound is supplied with analytical data and handling documentation for laboratory use.
  • Orally bioavailable TXNIP inhibitor.
  • Inhibits glucagon secretion and reduces hepatic glucose production.
  • Reverses hepatic steatosis in preclinical models.
  • High purity (99.16%) suitable for biochemical assays.
  • Soluble in DMSO and compatible with standard in vivo formulations.
  • Provided with SDS, COA, and handling instructions.
Catalog No. 50-002-21488