Learn More
TARGETMOL CHEMICALS INC SRI-37330 5MG
SDP
Catalog No. 502579706
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRI-37330 inhibits glucagon secretion and function reduces hepatic glucose production and reverses hepatic steatosis. purity: 99%
Catalog No. 50-257-9706 Supplier TARGETMOL CHEMICALS INC Supplier No. T600045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRI-37330 inhibits glucagon secretion and function reduces hepatic glucose production and reverses hepatic steatosis. purity: 99%