Learn More
Selleck Chemical LLC SRI-37330 HCl-E1272-5MG
SDP
Catalog No. 503138026
Encompass_Preferred
SRI-37330 HCl(EX-A5854) an anti-diabetic drug is an orally bioavailable inhibitor of thioredoxin-interacting protein (TXNIP) with an IC50 of 0 64 M for TXNIP expression in INS-1 cells Its treatment inhibits glucagon secretion and function decreases hepatic glucose production and reverses hepatic steatosis
Catalog No. 50-313-8026 Supplier Selleck Chemical LLC Supplier No. E12725MG
SRI-37330 HCl(EX-A5854) an anti-diabetic drug is an orally bioavailable inhibitor of thioredoxin-interacting protein (TXNIP) with an IC50 of 0 64 M for TXNIP expression in INS-1 cells Its treatment inhibits glucagon secretion and function decreases hepatic glucose production and reverses hepatic steatosis