Learn More
Medchemexpress LLC SRI-37330 hydrochloride | 2322245-49-6 | 99.7% | 424.87 g/mol | C16H20ClF3N4O2S | 5 MG
SDP
Catalog No. 5000223211
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor supplied as the hydrochloride salt for preclinical and biochemical research into metabolic disease and diabetes-related pathways. The compound is provided as a purified solid with solution formats available for assay use.
  • Orally bioavailable TXNIP inhibitor suitable for metabolic research.
  • Modulates glucagon secretion and reduces hepatic glucose production in models.
  • Supplied as a hydrochloride salt to aid solubility and handling.
  • High chemical purity (99.7%).
  • Available in small milligram quantities and solution formats for assays.
  • Documentation provided: datasheet and certificate of analysis.
Catalog No. 50-002-23211 Supplier Medchemexpress LLC Supplier No. HY1416235MG
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor supplied as the hydrochloride salt for preclinical and biochemical research into metabolic disease and diabetes-related pathways. The compound is provided as a purified solid with solution formats available for assay use.
  • Orally bioavailable TXNIP inhibitor suitable for metabolic research.
  • Modulates glucagon secretion and reduces hepatic glucose production in models.
  • Supplied as a hydrochloride salt to aid solubility and handling.
  • High chemical purity (99.7%).
  • Available in small milligram quantities and solution formats for assays.
  • Documentation provided: datasheet and certificate of analysis.