Learn More
Medchemexpress LLC SRI-37330 hydrochloride | 2322245-49-6 | 99.7% | C16H20ClF3N4O2S | 1 ML
SDP
Catalog No. 5000356935
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor. It reduces hepatic glucose production, reverses hepatic steatosis, and inhibits glucagon secretion and function, making it a promising compound for type 2 diabetes research.
  • Inhibits thioredoxin-interacting protein (TXNIP)
  • Reduces hepatic glucose production
  • Reverses hepatic steatosis
  • Potential for type 2 diabetes research
Catalog No. 50-003-56935 Supplier Medchemexpress LLC Supplier No. HY14162310MM/1ML
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor. It reduces hepatic glucose production, reverses hepatic steatosis, and inhibits glucagon secretion and function, making it a promising compound for type 2 diabetes research.
  • Inhibits thioredoxin-interacting protein (TXNIP)
  • Reduces hepatic glucose production
  • Reverses hepatic steatosis
  • Potential for type 2 diabetes research