Learn More
Medchemexpress LLC SRI-37330 (hydrochloride) | 2322245-49-6 | 99.7% | C16H20ClF3N4O2S | 10MG
SDP
Catalog No. 5000260699
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable small-molecule inhibitor of thioredoxin-interacting protein (TXNIP) used in preclinical metabolic research. It modulates glucagon secretion and signaling, reduces hepatic glucose production, and reverses hepatic steatosis, making it relevant for studies of type 2 diabetes and related metabolic disorders.
  • Orally bioavailable TXNIP inhibitor.
  • Modulates glucagon secretion and function.
  • Reduces hepatic glucose production.
  • Reverses hepatic steatosis in preclinical models.
  • High reported purity (99.72%).
  • Solid form suitable for storage and formulation.
  • Storage guidance: sealed at 4°C; in solvent, -80°C up to 6 months.
Catalog No. 50-002-60699 Supplier Medchemexpress LLC Supplier No. HY14162310MG
SRI-37330 hydrochloride is an orally bioavailable small-molecule inhibitor of thioredoxin-interacting protein (TXNIP) used in preclinical metabolic research. It modulates glucagon secretion and signaling, reduces hepatic glucose production, and reverses hepatic steatosis, making it relevant for studies of type 2 diabetes and related metabolic disorders.
  • Orally bioavailable TXNIP inhibitor.
  • Modulates glucagon secretion and function.
  • Reduces hepatic glucose production.
  • Reverses hepatic steatosis in preclinical models.
  • High reported purity (99.72%).
  • Solid form suitable for storage and formulation.
  • Storage guidance: sealed at 4°C; in solvent, -80°C up to 6 months.