Learn More
Medchemexpress LLC SRI-37330 hydrochloride | 2322245-49-6 | 99.7% | C16H20ClF3N4O2S | 25 MG
SDP
Catalog No. 5000352598
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor. It inhibits glucagon secretion and function, reduces hepatic glucose production, and reverses hepatic steatosis. This compound can be used for type 2 diabetes research.
  • Inhibits activity of human TXNIP promoter in INS-1 cells.
  • Decreases mRNA and protein levels of TXNIP in INS-1 cells.
  • Inhibits polymerase II (Pol II) binding to the E-box motif region of the TXNIP promoter.
  • Lowers glucagon secretion in TC1-6 cells.
  • Inhibits glucagon-induced glucose output from primary hepatocytes.
  • Decreases glucagon secretion and action in mice.
  • Blocks hepatic glucose output in mice.
  • Reverses obesity- and streptozotocin-induced diabetes and hepatic steatosis in mice.
  • Well tolerated in male C57BL/6J mice.
Catalog No. 50-003-52598 Supplier Medchemexpress LLC Supplier No. 25MGHY141623
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor. It inhibits glucagon secretion and function, reduces hepatic glucose production, and reverses hepatic steatosis. This compound can be used for type 2 diabetes research.
  • Inhibits activity of human TXNIP promoter in INS-1 cells.
  • Decreases mRNA and protein levels of TXNIP in INS-1 cells.
  • Inhibits polymerase II (Pol II) binding to the E-box motif region of the TXNIP promoter.
  • Lowers glucagon secretion in TC1-6 cells.
  • Inhibits glucagon-induced glucose output from primary hepatocytes.
  • Decreases glucagon secretion and action in mice.
  • Blocks hepatic glucose output in mice.
  • Reverses obesity- and streptozotocin-induced diabetes and hepatic steatosis in mice.
  • Well tolerated in male C57BL/6J mice.