Learn More
TARGETMOL CHEMICALS INC SRI 37892 5MG
SDP
Catalog No. 502578976
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRI 37892 is a Fzd7 inhibitor with potent activities against Wnt/(beta)-catenin signaling and cancer cell viability. purity: 98%
Catalog No. 50-257-8976 Supplier TARGETMOL CHEMICALS INC Supplier No. T96625MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRI 37892 is a Fzd7 inhibitor with potent activities against Wnt/(beta)-catenin signaling and cancer cell viability. purity: 98%