Learn More
Medchemexpress LLC SRI 6409-94 | 127697-58-9 | 99.25% | 379.49 | 5 MG
SDP
Catalog No. 5000359573
Encompass_Preferred
SRI 6409-94 is an orally active Ro 13-6298 analogue that is teratogenic. It serves as a molecular tool to study the effect of the three-dimensional configuration of retinol on teratogenic activity.
  • Orally active compound.
  • Aids in the study of retinol's three-dimensional configuration and its effect on teratogenic activity.
  • Induces deformed offspring in a dose-dependent manner in virgin female outbred Syrian golden hamsters.
  • Leads to significant reductions in mean fetal body weight and the number of ossified skeletal districts in studies.
  • Purity of 99.25%.
  • Available in solid form with a light yellow to brown color.
  • Stable for 3 years at -20°C as a powder, and 2 years at 4°C as a powder.
  • In solvent, stable for 6 months at -80°C and 1 month at -20°C.
Catalog No. 50-003-59573 Supplier Medchemexpress LLC Supplier No. HY1472515MG
SRI 6409-94 is an orally active Ro 13-6298 analogue that is teratogenic. It serves as a molecular tool to study the effect of the three-dimensional configuration of retinol on teratogenic activity.
  • Orally active compound.
  • Aids in the study of retinol's three-dimensional configuration and its effect on teratogenic activity.
  • Induces deformed offspring in a dose-dependent manner in virgin female outbred Syrian golden hamsters.
  • Leads to significant reductions in mean fetal body weight and the number of ossified skeletal districts in studies.
  • Purity of 99.25%.
  • Available in solid form with a light yellow to brown color.
  • Stable for 3 years at -20°C as a powder, and 2 years at 4°C as a powder.
  • In solvent, stable for 6 months at -80°C and 1 month at -20°C.