Learn More
Medchemexpress LLC SRI 6409-94 | 127697-58-9 | 99.3% | 379.49 | 1 ML
SDP
Catalog No. 5000362966
Encompass_Preferred
SRI 6409-94 is an orally active Ro 13-6298 analogue that is teratogenic. It serves as a molecular tool to study the effect of the three-dimensional configuration of retinol on teratogenic activity.
  • Causes virgin female outbred Syrian golden hamsters to produce deformed offspring in a dose-dependent manner.
  • Leads to significant reductions in mean fetal body weight and in the numbers of ossified skeletal districts.
Catalog No. 50-003-62966 Supplier Medchemexpress LLC Supplier No. HY14725110MM/1ML
SRI 6409-94 is an orally active Ro 13-6298 analogue that is teratogenic. It serves as a molecular tool to study the effect of the three-dimensional configuration of retinol on teratogenic activity.
  • Causes virgin female outbred Syrian golden hamsters to produce deformed offspring in a dose-dependent manner.
  • Leads to significant reductions in mean fetal body weight and in the numbers of ossified skeletal districts.