Learn More
Medchemexpress LLC Sri 6409-94 | 127697-58-9 | 99.3% | 379.49 | 25 MG
SDP
Catalog No. 5000363665
Encompass_Preferred
SRI 6409-94 is an orally active Ro 13-6298 analogue that is teratogenic. It serves as a molecular tool to study the effect of the three-dimensional configuration of retinol on teratogenic activity.
  • Orally active Ro 13-6298 analogue
  • Teratogenic
  • Molecular tool for studying retinol's three-dimensional configuration and teratogenic activity
  • Causes deformed offspring in a dose-dependent manner in virgin female outbred Syrian golden hamsters
  • Leads to significant reductions in mean fetal body weight and in the numbers of ossified skeletal districts
Catalog No. 50-003-63665 Supplier Medchemexpress LLC Supplier No. 25MGHY147251
SRI 6409-94 is an orally active Ro 13-6298 analogue that is teratogenic. It serves as a molecular tool to study the effect of the three-dimensional configuration of retinol on teratogenic activity.
  • Orally active Ro 13-6298 analogue
  • Teratogenic
  • Molecular tool for studying retinol's three-dimensional configuration and teratogenic activity
  • Causes deformed offspring in a dose-dependent manner in virgin female outbred Syrian golden hamsters
  • Leads to significant reductions in mean fetal body weight and in the numbers of ossified skeletal districts