Learn More
Aviva Systems Biology Corp SRI Antibody - C-terminal region (ARP62987_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62987_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRI. It was validated on Western Blot. . Alternate names: FLJ26259, SCN, CP22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-831