Learn More
Revvity Health Sciences Inc SRM-5 Small Rodent Manifold
Catalog No. 502016492
Encompass
SRM-5 small rodent manifold seamlessly delivers aneshesia to either 5 mice or 2 rats when used in conjunction with the Magnetic Quick Connect (sold separately). A robust new design requires no nose cones and offers superior scavenging for reduced anesthesia gas exposure. This system provides the ultimate in flexibility and ease of use as multiple accessories can be easily removed or changed with the quick connect functionality. For use with the IVIS Lumina S5 and IVIS Lumina X5 in vivo imaging systems.
Catalog No. 50-201-6492 Supplier Revvity Health Sciences Inc Supplier No. CLS148872
SRM-5 small rodent manifold seamlessly delivers aneshesia to either 5 mice or 2 rats when used in conjunction with the Magnetic Quick Connect (sold separately). A robust new design requires no nose cones and offers superior scavenging for reduced anesthesia gas exposure. This system provides the ultimate in flexibility and ease of use as multiple accessories can be easily removed or changed with the quick connect functionality. For use with the IVIS Lumina S5 and IVIS Lumina X5 in vivo imaging systems.