Learn More
Brammer Standard Company Inc SRM 1566B Oyster Tissue, Certified Reference Material, 25 g, Freeze-dried powder, Marine bivalve tissue
Catalog No. NC1426772
Encompass_Preferred
NIST SRM 1566b Oyster Tissue is a Certified Reference Material (CRM) used for the evaluation of analytical methods and instruments. This freeze-dried oyster tissue standard is designed to ensure the accuracy of mass fraction values for selected elements and proximates in marine bivalve tissue and similar matrices, offering a homogeneous and certified matrix for reliable analytical measurements.
  • Product type: Certified Reference Material (CRM)
  • Material format: Freeze-dried powder
  • Unit size: 25 g
  • Intended use: Evaluation of analytical methods and instruments
  • Application matrix: Marine bivalve tissue, foods, similar materials
  • Minimum sample mass: 250 mg (dry-mass basis)
  • Hazard classification: Not classified as a physical or health hazard
  • Certification validity: Until 01 June 2030
Catalog No. NC1426772 Supplier Brammer Standard Company Inc Supplier No. SRM 1566B
NIST SRM 1566b Oyster Tissue is a Certified Reference Material (CRM) used for the evaluation of analytical methods and instruments. This freeze-dried oyster tissue standard is designed to ensure the accuracy of mass fraction values for selected elements and proximates in marine bivalve tissue and similar matrices, offering a homogeneous and certified matrix for reliable analytical measurements.
  • Product type: Certified Reference Material (CRM)
  • Material format: Freeze-dried powder
  • Unit size: 25 g
  • Intended use: Evaluation of analytical methods and instruments
  • Application matrix: Marine bivalve tissue, foods, similar materials
  • Minimum sample mass: 250 mg (dry-mass basis)
  • Hazard classification: Not classified as a physical or health hazard
  • Certification validity: Until 01 June 2030