Learn More
Sino Biological Recombinant Human SIK1 Protein (GST Tag), Active
Catalog No. 502573136
Encompass_Preferred
Recombinant human SIK (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3136 Supplier Sino Biological Supplier No. S1411G500
Recombinant human SIK (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.