Learn More
Aviva Systems Biology Corp SRP14 Antibody - N-terminal region (ARP40454_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40454_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SRP14. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SRP14. Alternate names: ALURBPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53201