Learn More
Aviva Systems Biology Corp SRP14 Antibody - C-terminal region (ARP40453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRP14. It was validated on Western Blot. . Alternate names: ALURBP, MGC14326ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-872