Learn More
Aviva Systems Biology Corp SRP19 Antibody - middle region (ARP40456_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40456_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SRP19. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SRP19. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53202