Learn More
Aviva Systems Biology Corp SRP54A Antibody - C-terminal region (ARP40457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Srp54a. It was validated on Western Blot. . Alternate names: Srp54, Srp54aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-873