Learn More
Aviva Systems Biology Corp SRP68 Antibody - N-terminal region (ARP40757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRP68. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SRP68. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57614