Learn More
ABclonal Technology SRPK1 Rabbit mAb
SDP
Catalog No. 5000010301
Encompass_Preferred
This gene encodes a serine arginine protein kinase specific for the SR (serine arginine-rich domain) family of splicing factors The protein localizes to the nucleus and the cytoplasm It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors Alternative splicing of this gene results in multiple transcript variants Additional alternatively spliced transcript variants have been described for this gene but their full length nature have not been determined
Catalog No. 50-000-10301 Supplier ABclonal Technology Supplier No. A2598620UL
This gene encodes a serine arginine protein kinase specific for the SR (serine arginine-rich domain) family of splicing factors The protein localizes to the nucleus and the cytoplasm It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors Alternative splicing of this gene results in multiple transcript variants Additional alternatively spliced transcript variants have been described for this gene but their full length nature have not been determined