Learn More
Eurofins Discoverx SRPK2, active, 10 µg. (14-666)
Supplier:  Eurofins Discoverx 14666
Encompass
N-terminal GST-tagged recombinant, human SRPK2 amino acids 46-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9151