Learn More
Aviva Systems Biology Corp SRPRA Antibody - middle region (ARP40460_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40460_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRPR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SRPR. Alternate names: DP, MGC17355, MGC3650, MGC9571, SRP-alpha, SralphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-57462