Learn More
Aviva Systems Biology Corp SRPRB Antibody - middle region (ARP49643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRPRB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SRPRB. Alternate names: APMCF1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53593