Learn More
Aviva Systems Biology Corp Srpx Antibody - C-terminal region (ARP58956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Srpx. It was validated on Western Blot. . Alternate names: drs, drs-1, drs-2, mDRS-1, mDRS-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-540