Learn More
Aviva Systems Biology Corp Srrm1 Antibody - C-terminal region (ARP40607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Srrm1. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-884