Learn More
Aviva Systems Biology Corp SRRM4 Antibody - middle region (ARP60372_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60372_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRRM4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SRRM4. Alternate names: KIAA1853, MGC176729, MU-MB-2.76, nSR100ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-024