Learn More
Aviva Systems Biology Corp SRrp35 Antibody - middle region (ARP41089_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41089_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRrp35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SRrp35. Alternate names: FLJ14459, FLJ33484, FLJ41221, RP11-63L7.3, SFRS19, SRrp35, SFRS13BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57719