Learn More
Medchemexpress LLC HY-131387 25mg Medchemexpress, (S,R,S)-AHPC-Me-CO-CH2-PEG3-NH2 CAS:2409538-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13138725MG
Encompass_Preferred
Medchemexpress, HY-131387 25mg (S,R,S)-AHPC-Me-CO-CH2-PEG3-NH2 CAS:2409538-69-6 (S,R,S)-AHPC-Me-CO-CH2-PEG3-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the a VHL ligand and a linker. (S,R,S)-AHPC-Me-CO-CH2-PEG3-NH2 can be used in PROTAC BRD4 Degrader-5 (HY-133737) and PROTAC BRD4 Degrader-5-CO-PEG3-N3 (HY-133736). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1281