Learn More
TARGETMOL CHEMICALS INC SRS16-86 10MG
SDP
Catalog No. 502877040
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. SRS16-86 is a novel third-generation ferrostatin, is an inhibitor offerroptosis. Purity 99.85%
Catalog No. 50-287-7040 Supplier TARGETMOL CHEMICALS INC Supplier No. T907310MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. SRS16-86 is a novel third-generation ferrostatin, is an inhibitor offerroptosis. Purity 99.85%