Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Srt 1460 | 925432-73-1 | 98.2% | 507.60 | C26H29N5O4S | 10MG
SDP

Catalog No. 5000236711
Encompass_Preferred

SRT 1460 is a small-molecule activator of Sirtuin-1 (SIRT1) with an EC1.5 of 2.9 μM and strong selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM). It has been used in research to modulate SIRT1 activity and has been reported to be more potent than resveratrol in relevant assays.

  • Potent SIRT1 activation (EC1.5 2.9 μM).
  • High selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM).
  • High purity (~98.2%).
  • Molecular weight 507.60 g/mol.
  • Chemical formula C26H29N5O4S.
  • CAS number 925432-73-1.
  • Suitable for in vitro SIRT1 activation studies.
  • Available as a 10 mg research package.

Catalog No. 50-002-36711 Supplier Medchemexpress LLC Supplier No. HY12403710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SRT 1460 is a small-molecule activator of Sirtuin-1 (SIRT1) with an EC1.5 of 2.9 μM and strong selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM). It has been used in research to modulate SIRT1 activity and has been reported to be more potent than resveratrol in relevant assays.

  • Potent SIRT1 activation (EC1.5 2.9 μM).
  • High selectivity versus SIRT2 and SIRT3 (EC1.5 > 300 μM).
  • High purity (~98.2%).
  • Molecular weight 507.60 g/mol.
  • Chemical formula C26H29N5O4S.
  • CAS number 925432-73-1.
  • Suitable for in vitro SIRT1 activation studies.
  • Available as a 10 mg research package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.