Learn More
Medchemexpress LLC HY-10532 10mg Medchemexpress, SRT 1720 CAS:925434-55-5 Purity:>98%
SDP
Catalog No. 502080684
Encompass_Preferred
Medchemexpress, HY-10532 10mg SRT 1720 CAS:925434-55-5 SRT 1720 is a selective activator of human SIRT1 with an EC1.5 of 0.16 μM, and shows less potent activities agaiinst SIRT2 and SIRT3 with EC1.5s of 37 μM and > 300 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0684 Supplier Medchemexpress LLC Supplier No. HY1053210MG
Medchemexpress, HY-10532 10mg SRT 1720 CAS:925434-55-5 SRT 1720 is a selective activator of human SIRT1 with an EC1.5 of 0.16 μM, and shows less potent activities agaiinst SIRT2 and SIRT3 with EC1.5s of 37 μM and > 300 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.