Learn More
TARGETMOL CHEMICALS INC SRT 1720 25MG
SDP
Catalog No. 502579814
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRT 1720 is a selective activator of human SIRT1 (EC1.5 0.16 uM) and is >230-fold less potent for SIRT2 and SIRT3. purity: 99%
Catalog No. 50-257-9814 Supplier TARGETMOL CHEMICALS INC Supplier No. T509625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SRT 1720 is a selective activator of human SIRT1 (EC1.5 0.16 uM) and is >230-fold less potent for SIRT2 and SIRT3. purity: 99%