Learn More
Cayman Chemical SRT 1720 (hydrochloride), Purity: ≥98%, CAS: 1001645-58-4, 50 mg
SDP
Catalog No. 502888668
Encompass_Preferred
A SIRT1 activator (EC1.5 = 0.16 µM in a cell-free fluorescence polarization assay); reduces acetylated p53 levels in U2OS cells at 0.1 µM; reduces fed blood glucose levels, improves insulin sensitivity, and increases mitochondrial capacity in a DIO mouse model of diabetes at 100 mg/kg; improves liver, adipose tissue, and skeletal muscle insulin sensitivity in Zucker fa/fa rats; decreases synovitis and delays osteoarthritis progression in mice at 25 mg/kg
Catalog No. 50-288-8668 Supplier Cayman Chemical Supplier No. 1001102050 MG
A SIRT1 activator (EC1.5 = 0.16 µM in a cell-free fluorescence polarization assay); reduces acetylated p53 levels in U2OS cells at 0.1 µM; reduces fed blood glucose levels, improves insulin sensitivity, and increases mitochondrial capacity in a DIO mouse model of diabetes at 100 mg/kg; improves liver, adipose tissue, and skeletal muscle insulin sensitivity in Zucker fa/fa rats; decreases synovitis and delays osteoarthritis progression in mice at 25 mg/kg